Gene Transcripts

Gene Symbol
Tpbpb
Entrez Gene ID
116913
Gene Name
trophoblast specific protein beta
Gene Aliases
1600012N09Rik
Chromosome Location
Chr. 13: 60901295 - 60904847 on Build GRCm38
UniGene ID
Mm.443229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026429.4 NP_080705.1 3 201
GenBank mRNA AK005433.1 - 3 201
AK018793.1 - 3 200
AK018794.1 - 3 199
AK018795.1 - 3 201
AK018798.1 - 3 201
AK018799.1 - 3 201
AK018804.1 - 3 202
AK018805.1 - 3 201
AK018808.1 - 3 202
AK018809.1 - 3 200
AK018810.1 - 3 201
AK018820.1 - 3 201
AK018824.1 - 3 201
AK018827.1 - 3 200
AY034576.1 - 3 176
BC099441.1 - 3 186
BC100333.1 - 3 186 30787126

View Full Product Details