Gene Transcripts

Gene Symbol
Dclk2
Entrez Gene ID
70762
Gene Name
doublecortin-like kinase 2
Gene Aliases
6330415M09Rik, AU044875, CL2, CLICK2, Click-II, Dcamkl2
Chromosome Location
Chr. 3: 86786150 - 86920884 on Build GRCm38
UniGene ID
Mm.44490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195496.1 NP_001182425.1 1 978
NM_001195497.1 NP_001182426.1 1 978
NM_001195498.1 NP_001182427.1 1 978
NM_001195499.1 NP_001182428.1 1 978
NM_001195500.1 NP_001182429.1 1 978
NM_027539.5 NP_081815.3 1 978
GenBank mRNA AB198721.1 - 1 335
AK049126.1 - 1 468
AK049179.1 - 1 430
AK082633.1 - 1 474
AY968049.1 - 1 266
AY968050.1 - 1 266
AY968051.1 - 1 266
BC056921.1 - 1 946 6822681
BC060192.1 - 1 282

View Full Product Details