Gene Transcripts

Gene Symbol
Hmha1
Entrez Gene ID
70719
Gene Name
histocompatibility (minor) HA-1
Gene Aliases
6330406L22Rik, AW539505, Ha-1
Chromosome Location
Chr. 10: 80016672 - 80031471 on Build GRCm38
UniGene ID
Mm.243954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142701.1 NP_001136173.1 10 1305
NM_027521.3 NP_081797.1 9 857
GenBank mRNA AK044860.1 - 9 819
AK134029.1 - 3 279
AK137810.1 - 1 2979
AK151423.1 - 10 1249
AK159194.1 - 10 1306
AK159474.1 - 9 1349
AK161766.1 - 2 365
AK170262.1 - 10 1291
AK171304.1 - 10 1291
BC053750.1 - 9 857 6333971

View Full Product Details