Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031197.2 | NP_112474.2 | 7, 8 | 1071 | |
GenBank mRNA | AK005068.1 | - | 7, 8 | 1072 | |
AK165430.1 | - | 7, 8 | 1006 | ||
BC034675.1 | - | 7, 8 | 990 | 4236331 | |
X15684.1 | - | 7, 8 | 1020 | ||
X16986.1 | - | 7, 8 | 1010 |