Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033541.4 | NP_291019.1 | 7 | 1810 | |
GenBank mRNA | AB067528.1 | - | 6 | 1467 | |
AF459815.1 | - | 6 | 1467 | ||
AF459816.1 | - | 6 | 1467 | ||
AK028986.1 | - | 7 | 1811 | ||
AK078952.1 | - | 6 | 1480 | ||
AK163253.1 | - | 6 | 1483 |