Gene Transcripts

Gene Symbol
Peli2
Entrez Gene ID
93834
Gene Name
pellino 2
Gene Aliases
AW047589, BB129927
Chromosome Location
Chr. 14: 48120778 - 48308103 on Build GRCm38
UniGene ID
Mm.296457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033602.2 NP_291080.2 3 519
GenBank mRNA AF302504.1 - 3 448
AK030564.1 - 3 566
AK045515.1 - 2 392
BC027062.1 - 2 358 5355240

View Full Product Details