Gene Transcripts

Gene Symbol
Fxyd4
Entrez Gene ID
108017
Gene Name
FXYD domain-containing ion transport regulator 4
Gene Aliases
0610008I02Rik, AI267073, Chif
Chromosome Location
Chr. 6: 117933559 - 117938510 on Build GRCm38
UniGene ID
Mm.250392
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173372.1 NP_001166843.1 9 414
NM_033648.1 NP_387468.1 9 417
GenBank mRNA AK018728.1 - 9 414
BC086918.1 - 10 678 6762406

View Full Product Details