Gene Transcripts

Gene Symbol
Epgn
Entrez Gene ID
71920
Gene Name
epithelial mitogen
Gene Aliases
2310069M11Rik, epigen
Chromosome Location
Chr. 5: 91027517 - 91035212 on Build GRCm38
UniGene ID
Mm.197319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053087.2 NP_444317.1 5 729
GenBank mRNA AJ291391.1 - 5 729
AK010136.1 - 4 677

View Full Product Details