Gene Transcripts

Gene Symbol
Hemgn
Entrez Gene ID
93966
Gene Name
hemogen
Gene Aliases
4921524M03Rik, AI317176, EDAG, Hgn, mNDR
Chromosome Location
Chr. 4: 46393989 - 46413504 on Build GRCm38
UniGene ID
Mm.25793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053149.2 NP_444379.1 2 347
GenBank mRNA AF269248.1 - 2 347
AK006588.1 - 3 362
AK076587.1 - 3 332
AK077692.1 - 2 190
AK156536.1 - 2 275
AK171890.1 - 2 266
AK171906.1 - 2 396
AK172421.1 - 2 266
AY699986.1 - 2 189
BC050798.1 - 3 326 6743819

View Full Product Details