Gene Transcripts
- Gene Symbol
- Vmn1r12
- Entrez Gene ID
- 626397
- Gene Name
- vomeronasal 1 receptor 12
- Gene Aliases
- EG626397, Gm6674
- Chromosome Location
- Chr. 6: 57158920 - 57159843 on Build GRCm38
- UniGene ID
- Mm.475092
- Species
- Mus musculus
- Gene Symbol
- Vmn1r14
- Entrez Gene ID
- 113864
- Gene Name
- vomeronasal 1 receptor 14
- Gene Aliases
- V1rc7
- Chromosome Location
- Chr. 6: 57233439 - 57234350 on Build GRCm38
- UniGene ID
- Mm.431974
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053237.2 |
NP_444467.2
|
1 |
577 |
|
View Full Product Details