Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146324.1 | NP_001139796.1 | 2 | 337 | |
NM_080634.4 | NP_542365.3 | 2 | 337 | ||
GenBank mRNA | AF393747.1 | - | 2 | 326 | |
AF393780.1 | - | 2 | 329 | ||
AK034377.1 | - | 2 | 308 | ||
AK036663.1 | - | 2 | 345 | ||
AK049204.1 | - | 2 | 330 | ||
AK079896.1 | - | 2 | 329 | ||
AK086167.1 | - | 2 | 324 | ||
AK155497.1 | - | 2 | 304 | ||
AK157953.1 | - | 2 | 322 | ||
AK166194.1 | - | 2 | 322 | ||
AK169855.1 | - | 2 | 308 | ||
BC058235.1 | - | 2 | 311 | 6307400 |