Gene Transcripts

Gene Symbol
Cyp4f13
Entrez Gene ID
170716
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 13
Gene Aliases
0610030I10Rik, Cypf13
Chromosome Location
Chr. 17: 32924688 - 32947415 on Build GRCm38
UniGene ID
Mm.254838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130882.1 NP_570952.1 7 925
GenBank mRNA AF233643.1 - 7 925
BC003954.1 - 7 934 3487830
BC058330.1 - 4 617

View Full Product Details