Gene Transcripts
- Gene Symbol
- ORM2
- Entrez Gene ID
- 5005
- Gene Name
- orosomucoid 2
- Gene Aliases
- AGP-B, AGP-B', AGP2
- Chromosome Location
- Chr. 9: 114329789 - 114333256 on Build GRCh38
- UniGene ID
- Hs.719954
- Species
- Homo sapiens
- Gene Symbol
- ORM1
- Entrez Gene ID
- 5004
- Gene Name
- orosomucoid 1
- Gene Aliases
- AGP-A, AGP1, HEL-S-153w, ORM
- Chromosome Location
- Chr. 9: 114323023 - 114326479 on Build GRCh38
- UniGene ID
- Hs.522356
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC143314.1 |
- |
5 |
542 |
|
View Full Product Details