Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145510.1 | NP_663485.1 | 2 | 1552 | |
GenBank mRNA | AK087547.1 | - | 2 | 1547 | |
AK155054.1 | - | 2 | 1552 | ||
AK156126.1 | - | 1464 | |||
AK158129.1 | - | 1565 | |||
BC011166.1 | - | 2 | 1552 | 4206804 | |
BC024808.1 | - | 2 | 1542 | 5356768 |