Gene Transcripts

Gene Symbol
Plekhh3
Entrez Gene ID
217198
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 3
Gene Aliases
BC020025
Chromosome Location
Chr. 11: 101162679 - 101171405 on Build GRCm38
UniGene ID
Mm.273254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146030.2 NP_666142.1 3 700
GenBank mRNA AK154510.1 - 3 716
AK164128.1 - 3 716
BC020025.1 - 3 700 3490992

View Full Product Details