Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
5891
Gene Name
MOK protein kinase
Gene Aliases
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location
Chr. 14: 102224450 - 102305200 on Build GRCh38
UniGene ID
Hs.104119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272011.1 NP_001258940.1 8 879
NM_014226.2 NP_055041.1 9 969
NR_073540.1 3 317
NR_073541.1 4 364
NR_073542.1 3 317
XM_006720225.2 XP_006720288.1 9 969
XM_006720226.2 XP_006720289.1 9 969
XM_011537052.1 XP_011535354.1 9 969
XM_011537053.2 XP_011535355.1 3 996
XM_017021549.1 XP_016877038.1 9 966
XM_017021550.1 XP_016877039.1 8 865
XM_017021551.1 XP_016877040.1 10 909
XM_017021552.1 XP_016877041.1 9 1007
XM_017021553.1 XP_016877042.1 7 804
XM_017021554.1 XP_016877043.1 7 803
XM_017021555.1 XP_016877044.1 9 966
XM_017021557.1 XP_016877046.1 8 869
XM_017021558.1 XP_016877047.1 8 832
XM_017021559.1 XP_016877048.1 8 958
GenBank mRNA AB022694.1 - 9 963
AK094747.1 - 6 1755
AK131542.1 - 1 62
AK302349.1 - 8 864
AK312778.1 - 9 960
BC026069.1 - 8 1380 4838296
BC043179.1 - 7 1725 5287879
BC053536.1 - 8 1924 5175851
U46192.1 - 3 307
U46193.1 - 4 364
U46194.1 - 3 303

View Full Product Details