Gene Transcripts

Gene Symbol
Pop1
Entrez Gene ID
67724
Gene Name
processing of precursor 1, ribonuclease P/MRP family, (S. cerevisiae)
Gene Aliases
4932434G09Rik
Chromosome Location
Chr. 15: 34495311 - 34530653 on Build GRCm38
UniGene ID
Mm.248779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026340.3 NP_080616.1 5, 6 850
NM_152894.2 NP_690854.1 5, 6 850
GenBank mRNA AK016547.1 - 5, 6 837
BC034742.1 - 5, 6 829 4935194

View Full Product Details