Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_170593.3 | NP_733481.1 | 5 | 753 | |
GenBank mRNA | AK035975.1 | - | 2 | 1442 | |
AK036480.1 | - | 734 | |||
AK045560.1 | - | 754 | |||
AY150699.1 | - | 5 | 554 | ||
BC118526.1 | - | 5 | 548 | 40099214 |