Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145958.1 | NP_001139430.1 | 15 | 2090 | |
NM_172122.2 | NP_742120.2 | 16 | 2343 | ||
GenBank mRNA | AF527975.1 | - | 16 | 2343 | |
AK122285.1 | - | 8 | 1093 | ||
BC036729.1 | - | 1 | 81 | ||
BC054054.1 | - | 14 | 2071 | 6394697 | |
CA749493.1 | - | 1 | 14 |