Gene Transcripts
- Gene Symbol
- Dnajc11
- Entrez Gene ID
- 230935
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 11
- Gene Aliases
- E030019A03Rik
- Chromosome Location
- Chr. 4: 151933720 - 151981959 on Build GRCm38
- UniGene ID
- Mm.21353
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172704.3 |
NP_766292.2
|
2, 3 |
233 |
|
GenBank mRNA |
AK053156.1 |
- |
2, 3 |
215 |
|
AK089983.1 |
- |
2, 3 |
225 |
|
AK156793.1 |
- |
2, 3 |
215 |
|
AK171781.1 |
- |
2, 3 |
214 |
|
AK172542.1 |
- |
2, 3 |
214 |
|
BC075619.1 |
- |
2, 3 |
209 |
|
BC079852.1 |
- |
2, 3 |
203 |
|
BC085257.1 |
- |
2, 3 |
207 |
5414963 |
BY784439.1 |
- |
2, 3 |
244 |
|
View Full Product Details