Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172827.3 | NP_766415.1 | 2 | 289 | |
GenBank mRNA | AK029094.1 | - | 2 | 290 | |
AK036967.1 | - | 2 | 290 | ||
BC066791.1 | - | 3 | 613 | 30054751 | |
BC120925.1 | - | 2 | 246 | 40098425 | |
BC120926.1 | - | 2 | 246 | 40098428 |