Gene Transcripts

Gene Symbol
Eny2
Entrez Gene ID
223527
Gene Name
enhancer of yellow 2 homolog (Drosophila)
Gene Aliases
1810057B09Rik, 6720481I12, DC6, Ey2
Chromosome Location
Chr. 15: 44428081 - 44438256 on Build GRCm38
UniGene ID
Mm.291828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175009.3 NP_778174.1 5 2166
GenBank mRNA AK032957.1 - 5 2166
AK136334.1 - 1 1253
AK162511.1 - 5 2161

View Full Product Details