Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164491.1 | NP_001157963.1 | 2 | 199 | |
NM_198649.3 | NP_941051.2 | 1 | 518 | ||
GenBank mRNA | AK082380.1 | - | 1 | 517 | |
AK086538.1 | - | 1 | 298 | ||
AK173040.1 | - | 1 | 76 | ||
BC060275.1 | - | 2 | 170 | 6822626 | |
BC094229.1 | - | 1 | 492 | 30534236 |