Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001398.2 | NP_001389.2 | 2 | 326 | |
XM_017026448.1 | XP_016881937.1 | 2 | 276 | ||
GenBank mRNA | AK291860.1 | - | 2 | 261 | |
AK301211.1 | - | 2 | 258 | ||
BC011792.2 | - | 2 | 243 | 4300082 | |
BC017408.1 | - | 2 | 267 | 4306900 | |
BG527600.1 | - | 2 | 327 | ||
U16660.1 | - | 2 | 268 |