Gene Transcripts
- Gene Symbol
- Nfxl1
- Entrez Gene ID
- 100978
- Gene Name
- nuclear transcription factor, X-box binding-like 1
- Gene Aliases
- 1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
- Chromosome Location
- Chr. 5: 72513301 - 72559707 on Build GRCm38
- UniGene ID
- Mm.187453
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK051057.1 |
- |
1 |
1380 |
|
AK085068.1 |
- |
1 |
1375 |
|
AK164247.1 |
- |
1 |
1994 |
|
View Full Product Details