Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP8
Entrez Gene ID
838177
Gene Name
tubby like protein 8
Gene Aliases
AT1G16070, AtTLP8, T24D18.17, T24D18_17, tubby like protein 8
Chromosome Location
Chr.1: 5510301 - 5513980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332209.1 NP_001319017.1 7 - 8 1385 105
NM_001332210.1 NP_001322582.1 6 - 7 1440 105
NM_101475.3 NP_173059.1 8 - 9 1318 105
GenBank mRNA AF487269.1 - 8 - 9 1117 105
BT025624.1 - 8 - 9 1117 105

View Full Product Details