Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSH8
Entrez Gene ID
838262
Gene Name
LIGHT-DEPENDENT SHORT HYPOCOTYLS-like protein (DUF640)
Gene Aliases
AT1G16910, F17F16.11, LIGHT SENSITIVE HYPOCOTYLS 8
Chromosome Location
Chr.1: 5785003 - 5785959 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101552.3 NP_173135.1 1 - 1 428 84
GenBank mRNA AY800583.1 - 1 - 1 428 84

View Full Product Details