Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRG1
Entrez Gene ID
838320
Gene Name
developmentally regulated G-protein 1
Gene Aliases
AT1G17470, ATDRG, ATDRG1, F28G4.4, developmentally regulated G-protein 1
Chromosome Location
Chr.1: 6002888 - 6006410 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084086.1 NP_001077555.1 3 - 4 217 92
NM_101609.4 NP_173190.1 3 - 4 292 92
GenBank mRNA BT021092.1 - 2 - 3 155 92

View Full Product Details