The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_101721.4 | NP_173298.1 | 4 - 5 | 521 | 130 |
GenBank mRNA | AK227482.1 | - | 4 - 5 | 407 | 130 |
BT005999.1 | - | 3 - 4 | 298 | 130 |