Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SPL14
- Entrez Gene ID
-
838692
- Gene Name
- squamosa promoter binding protein-like 14
- Gene Aliases
-
AT1G20980, ATSPL14, F9H16.3, F9H16_3, FBR6, SPL1R2, SQUAMOSA PROMOTER BINDING PROTEIN-LIKE 14, squamosa promoter binding protein-like 14
- Chromosome Location
-
Chr.1: 7324471 - 7329441 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101951.4 |
NP_173522.1 |
5 - 6 |
2259 |
86 |
View Full Product Details