Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAK2
Entrez Gene ID
838723
Gene Name
wall-associated kinase 2
Gene Aliases
AT1G21270, F16F4.5, F16F4_5, wall-associated kinase 2
Chromosome Location
Chr.1: 7444749 - 7448483 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101979.4 NP_173549.1 1 - 2 1074 119
GenBank mRNA AY062531.1 - 1 - 2 892 119
BT010335.1 - 1 - 2 826 119

View Full Product Details