Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YUC11
Entrez Gene ID
838741
Gene Name
Flavin-binding monooxygenase family protein
Gene Aliases
AT1G21430, F24J8.6, F24J8_6
Chromosome Location
Chr.1: 7500845 - 7502186 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101994.1 NP_173564.1 1 - 2 622 76

View Full Product Details