Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOLT1
Entrez Gene ID
836770
Gene Name
folate transporter 1
Gene Aliases
AT5G66380, ATFOLT1, K1F13.3, K1F13_3, folate transporter 1
Chromosome Location
Chr.5: 26513262 - 26515800 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126036.4 NP_569032.1 2 - 3 449 63
GenBank mRNA AK227464.1 - 2 - 3 314 63
BT010139.1 - 2 - 3 182 63

View Full Product Details