Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABA1
- Entrez Gene ID
-
836838
- Gene Name
- zeaxanthin epoxidase (ZEP) (ABA1)
- Gene Aliases
-
ABA DEFICIENT 1, ARABIDOPSIS THALIANA ABA DEFICIENT 1, ARABIDOPSIS THALIANA ZEAXANTHIN EPOXIDASE, AT5G67030, ATABA1, ATZEP, IBS3, IMPAIRED IN BABA-INDUCED STERILITY 3, K8A10.10, K8A10_10, LOS6, LOW EXPRESSION OF OSMOTIC STRESS-RESPONSIVE GENES 6, NON-PHOTOCHEMICAL QUENCHING 2, NPQ2, ZEAXANTHIN EPOXIDASE, ZEP
- Chromosome Location
-
Chr.5: 26753469 - 26757402 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_126103.2 |
NP_201504.2 |
13 - 14 |
1901 |
102 |
View Full Product Details