Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABA1
Entrez Gene ID
836838
Gene Name
zeaxanthin epoxidase (ZEP) (ABA1)
Gene Aliases
ABA DEFICIENT 1, ARABIDOPSIS THALIANA ABA DEFICIENT 1, ARABIDOPSIS THALIANA ZEAXANTHIN EPOXIDASE, AT5G67030, ATABA1, ATZEP, IBS3, IMPAIRED IN BABA-INDUCED STERILITY 3, K8A10.10, K8A10_10, LOS6, LOW EXPRESSION OF OSMOTIC STRESS-RESPONSIVE GENES 6, NON-PHOTOCHEMICAL QUENCHING 2, NPQ2, ZEAXANTHIN EPOXIDASE, ZEP
Chromosome Location
Chr.5: 26753469 - 26757402 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126103.2 NP_201504.2 8 - 9 1522 98
NM_180954.3 NP_851285.1 8 - 9 1688 98
GenBank mRNA AY093145.1 - 8 - 9 1456 98

View Full Product Details