Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDN3
Entrez Gene ID
836859
Gene Name
small RNA degrading nuclease 3
Gene Aliases
AT5G67240, K21H1.20, K21H1_20, small RNA degrading nuclease 3
Chromosome Location
Chr.5: 26824236 - 26828318 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345817.1 NP_001318895.1 9 - 10 1187 69
NM_001345818.1 NP_001331564.1 9 - 10 1211 69
NM_001345819.1 NP_001331563.1 9 - 10 1187 69
NM_126124.4 NP_201525.2 9 - 10 1299 69
GenBank mRNA AY080843.1 - 9 - 10 1150 69
BT001977.1 - 9 - 10 1079 69

View Full Product Details