Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRB2
Entrez Gene ID
836894
Gene Name
Homeodomain-like/winged-helix DNA-binding family protein
Gene Aliases
AT5G67580, ATTBP3, ATTRB2, K9I9.15, K9I9_15, TBP3, TELOMERE REPEAT BINDING FACTOR 2, TELOMERE-BINDING PROTEIN 3
Chromosome Location
Chr.5: 26955568 - 26957774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126158.3 NP_201559.1 2 - 3 347 112
NM_180955.3 NP_851286.1 2 - 3 437 112
GenBank mRNA AY039883.1 - 2 - 3 342 112
AY077668.1 - 1 - 2 152 112
AY519545.1 - 1 - 2 152 112

View Full Product Details