Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHA2B
Entrez Gene ID
814644
Gene Name
RING-H2 finger protein 2B
Gene Aliases
AT2G01150, F10A8.3, F10A8_3, RING-H2 FINGER PROTEIN RHA2B, RING-H2 finger protein 2B
Chromosome Location
Chr.2: 100484 - 101566 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335035.1 NP_001318169.1 1 - 1 743 94
NM_001335036.1 NP_001324450.1 1 - 1 893 94
NM_126177.3 NP_565253.2 1 - 1 814 94

View Full Product Details