Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR14
Entrez Gene ID
814707
Gene Name
response regulator 14
Gene Aliases
ARR14, AT2G01760, T8O11.7, T8O11_7, response regulator 14
Chromosome Location
Chr.2: 332902 - 334760 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126237.3 NP_178285.1 1 - 2 230 135
GenBank mRNA AY099726.1 - 1 - 2 180 135
AY128887.1 - 1 - 2 106 135

View Full Product Details