Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MBD9
- Entrez Gene ID
-
821132
- Gene Name
- methyl-CPG-binding domain 9
- Gene Aliases
-
AT3G01460, ATMBD9, T13O15.10, T13O15_10, methyl-CPG-binding domain 9
- Chromosome Location
-
Chr.3: 173190 - 182454 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111012.4 |
NP_186795.1 |
4 - 5 |
3128 |
71 |
View Full Product Details