Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPLATE
Entrez Gene ID
821081
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT3G01780, F28J7.11, F28J7_11
Chromosome Location
Chr.3: 278882 - 283664 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337358.1 NP_001327599.1 4 - 5 2597 117
NM_111044.4 NP_186827.2 4 - 5 2684 117

View Full Product Details