Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KUP3
Entrez Gene ID
821063
Gene Name
K+ uptake transporter 3
Gene Aliases
ARABIDOPSIS THALIANA POTASSIUM TRANSPORTER 4, AT3G02050, ATKT4, ATKUP3, F1C9.17, F1C9_17, K+ uptake transporter 3, KUP3P
Chromosome Location
Chr.3: 350613 - 354345 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111071.3 NP_186854.1 6 - 7 1099 106
GenBank mRNA AY062747.1 - 6 - 7 1096 106
BT003375.1 - 6 - 7 897 106

View Full Product Details