Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPM4
Entrez Gene ID
821170
Gene Name
BTB-POZ and MATH domain 4
Gene Aliases
AT3G03740, ATBPM4, BTB-POZ AND MATH DOMAIN 4, BTB-POZ and MATH domain 4, F20H23.23, F20H23_23
Chromosome Location
Chr.3: 936878 - 941670 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111245.3 NP_566212.2 1 - 2 2318 132
GenBank mRNA AF367309.1 - 1 - 2 456 132
AY059150.1 - 1 - 2 455 132

View Full Product Details