Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC14
Entrez Gene ID
827759
Gene Name
plastid transcriptionally active 14
Gene Aliases
AT4G20130, F1C12.4, TAC14, plastid transcriptionally active 14
Chromosome Location
Chr.4: 10878817 - 10882052 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341400.1 NP_001327970.1 4 - 5 611 111
NM_001341401.1 NP_001327969.1 2 - 3 291 111
NM_118132.5 NP_193746.3 5 - 6 704 111
GenBank mRNA AK230206.1 - 5 - 6 626 111
BT004184.1 - 5 - 6 618 111
BT005475.1 - 5 - 6 607 111

View Full Product Details