Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMN/FHY
Entrez Gene ID
828232
Gene Name
riboflavin kinase/FMN hydrolase
Gene Aliases
AT4G21470, ATFMN/FHY, F18E5.90, F18E5_90, riboflavin kinase/FMN hydrolase
Chromosome Location
Chr.4: 11431104 - 11433322 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118267.5 NP_193878.2 9 - 10 1200 105
GenBank mRNA AK227237.1 - 9 - 10 1181 105
AY878327.1 - 9 - 10 1020 105
BT006373.1 - 9 - 10 1020 105

View Full Product Details