Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDB3
Entrez Gene ID
828234
Gene Name
NAD(P)H dehydrogenase B3
Gene Aliases
AT4G21490, F18E5.110, F18E5_110, NAD(P)H dehydrogenase B3
Chromosome Location
Chr.4: 11436326 - 11439506 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118269.6 NP_193880.5 2 - 3 462 116

View Full Product Details