Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GA3OX3
Entrez Gene ID
828256
Gene Name
gibberellin 3-oxidase 3
Gene Aliases
ARABIDOPSIS THALIANA GIBBERELLIN 3-OXIDASE 3, AT4G21690, ATGA3OX3, F17L22.150, F17L22_150, gibberellin 3-oxidase 3
Chromosome Location
Chr.4: 11527229 - 11529060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118289.1 NP_193900.1 1 - 2 463 105
GenBank mRNA DQ446858.1 - 1 - 2 463 105

View Full Product Details