Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP2A
Entrez Gene ID
828349
Gene Name
OSBP(oxysterol binding protein)-related protein 2A
Gene Aliases
AT4G22540, F7K2.120, F7K2_120, OSBP(oxysterol binding protein)-related protein 2A
Chromosome Location
Chr.4: 11860409 - 11866475 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036621.3 NP_001031698.1 3 - 4 552 104
NM_001203871.1 NP_001190800.1 3 - 4 563 104
NM_001341552.1 NP_001328217.1 3 - 4 1166 104
NM_001341553.1 NP_001328218.1 3 - 4 977 104
NM_118380.3 NP_567662.1 3 - 4 1200 104
NM_202863.3 NP_974592.1 3 - 4 566 104
GenBank mRNA AK229811.1 - 3 - 4 530 104
AY052361.1 - 3 - 4 1085 104
BT001093.1 - 3 - 4 833 104

View Full Product Details