Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGT1A
Entrez Gene ID
828457
Gene Name
phosphatase-like protein
Gene Aliases
AT4G23570, F9D16.40, F9D16_40
Chromosome Location
Chr.4: 12299749 - 12302935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036627.2 NP_001031704.1 8 - 9 982 72
NM_118488.6 NP_194088.1 9 - 10 1024 72
NM_179098.3 NP_849429.1 8 - 9 1108 72
GenBank mRNA AY063978.1 - 8 - 9 979 72
AY096395.1 - 8 - 9 842 72

View Full Product Details