Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPT1
Entrez Gene ID
828466
Gene Name
polyprenyltransferase 1
Gene Aliases
4-HYDROXYBENZOATE POLYPRENYL DIPHOSPHATE TRANSFERASE, AT4G23660, AtPPT1, F9D16.130, F9D16_130, polyprenyltransferase 1
Chromosome Location
Chr.4: 12327638 - 12331876 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203886.1 NP_001190815.1 4 - 5 761 113
NM_001341624.1 NP_001320050.1 3 - 4 1078 113
NM_001341625.1 NP_001328939.1 4 - 5 819 113
NM_118497.4 NP_567688.1 3 - 4 1100 113
NM_179100.4 NP_849431.1 4 - 5 846 113
GenBank mRNA AB052553.2 - 3 - 4 898 113
AY059864.1 - 4 - 5 745 113
BT008496.1 - 3 - 4 700 113

View Full Product Details